APO455830268 Apollo Scientific Ltd

1-(4-Chloro-2-nitrophenyl)piperazine

  • MDL number MFCD00640777
  • Cas Number : 405910-34-1
  • Molecular Weight : 241.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Chloro-2-(piperazin-1-yl)nitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-chloro-2-nitrophenylpiperazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-chloro-2-nitrophenylpiperazine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart