APO455830269 Apollo Scientific Ltd

3-(tert-Butyl)aniline hydrochloride

  • MDL number MFCD08273794
  • Cas Number : 83672-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 110/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32859

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32859
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart