APO455830280 Apollo Scientific Ltd

2-Bromo-3-methyl-5-nitropyridine

  • MDL number MFCD03095065
  • Cas Number : 23132-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-59
Synonym 2-Bromo-5-nitro-3-picoline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-methyl-5-nitropyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-methyl-5-nitropyridine
pictograms GHS07
hstatements H302+H312;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart