APO455830281 Apollo Scientific Ltd

5-Bromopyrimidine-2-carbonitrile

  • MDL number MFCD02940446
  • Cas Number : 38275-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Synonym 5-Bromo-2-cyanopyrimidine, 5-Bromo-2-cyano-1,3-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromopyrimidine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromopyrimidine-2-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart