APO455830283 Apollo Scientific Ltd

N'-Cyanopyridine-4-carboxamidine

  • MDL number MFCD09702431
  • Cas Number : 23275-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N'-Cyanopyridine-4-carboximidamide, N'-Cyanoisonicotinamidine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36160

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36160
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart