APO455830284 Apollo Scientific Ltd

Methyl (2-hydroxybenzoyl)imidoformate

  • MDL number MFCD09998770
  • Cas Number : 1000018-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(Methoxymethylene)salicylamide, Methyl N-(2-hydroxybenzoyl)methanimidate
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36162

Safety Information

signalword Danger
pcodes P280;P301+P310;P302+P352
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36162
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart