APO455830286 Apollo Scientific Ltd

3-Bromo-2-furaldehyde

  • MDL number MFCD07368224
  • Cas Number : 14757-78-9
  • Molecular Weight : 174.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-2-formylfuran, 3-Bromofuran-2-carboxaldehyde
Purity 96
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-furaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-furaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart