APO455830291 Apollo Scientific Ltd

3-Bromopyridine-2-carboxaldehyde

  • MDL number MFCD07781220
  • Cas Number : 405174-97-2
  • Molecular Weight : 186.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Synonym 3-Bromo-2-formylpyridine, 3-Bromopicolinaldehyde
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromopyridine-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromopyridine-2-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart