APO455830294 Apollo Scientific Ltd

2-Isopropyl-1H-benzimidazole

  • MDL number MFCD00051759
  • Cas Number : 5851-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-239
Synonym 2-(Prop-2-yl)-1H-benzimidazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14995

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14995
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart