APO455830296 Apollo Scientific Ltd

2-Bromo-3-hydroxybenzaldehyde

  • MDL number MFCD00092081
  • Cas Number : 196081-71-7
  • Molecular Weight : 201.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-3-hydroxybenzaldehyde
Inchi Code 1S/C7H5BrO2/c8-7-5(4-9)2-1-3-6(7)10/h1-4,10H
Inchi Key OHXPHMPERMIICA-UHFFFAOYSA-N

Description

Synonym 2-Bromo-3-formylphenol
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-hydroxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-hydroxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart