APO455830297 Apollo Scientific Ltd

2,4-Dichloro-1,3,5-triazine

  • MDL number MFCD04115347
  • Cas Number : 2831-66-5
  • Molecular Weight : 149.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 50-56
Iupac Name 2,4-dichloro-1,3,5-triazine
Inchi Code 1S/C3HCl2N3/c4-2-6-1-7-3(5)8-2/h1H
Inchi Key OMRXVBREYFZQHU-UHFFFAOYSA-N

Description

Synonym 2,4-Dichloro-s-triazine
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 50-56
Product Link https://store.apolloscientific.co.uk/product/24-dichloro-135-triazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-dichloro-135-triazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart