APO455830299 Apollo Scientific Ltd

3-Bromo-5-(hydroxymethyl)pyridine

  • MDL number MFCD03265757
  • Cas Number : 37669-64-0
  • Molecular Weight : 188.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromo-3-pyridinyl)methanol
Inchi Code 1S/C6H6BrNO/c7-6-1-5(4-9)2-8-3-6/h1-3,9H,4H2
Inchi Key WDVDHJLKXYCOFS-UHFFFAOYSA-N

Description

Synonym (5-Bromopyridin-3-yl)methanol
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-hydroxymethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-hydroxymethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart