APO455830302 Apollo Scientific Ltd

trans-4-Aminocyclohexan-1-ol, N-BOC protected

  • MDL number MFCD03844613
  • Cas Number : 111300-06-2
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 169-173
Iupac Name tert-butyl 4-hydroxycyclohexylcarbamate
Inchi Code 1S/C11H21NO3/c1-11(2,3)15-10(14)12-8-4-6-9(13)7-5-8/h8-9,13H,4-7H2,1-3H3,(H,12,14)/t8-,9-
Inchi Key DQARDWKWPIRJEH-KYZUINATSA-N

Description

Synonym tert-Butyl (trans-4-hydroxycyclohex-1-yl)carbamate, trans-4-[(tert-Butoxycarbonyl)amino]cyclohexan-1-ol
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 169-173
Product Link https://store.apolloscientific.co.uk/product/trans-4-aminocyclohexan-1-ol-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/trans-4-aminocyclohexan-1-ol-n-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart