APO455830310 Apollo Scientific Ltd

Imidazo[1,2-a]pyridine-6-boronic acid

  • MDL number MFCD06739225
  • Cas Number : 913835-63-9
  • Molecular Weight : 161.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 115-117
Iupac Name imidazo[1,2-a]pyridin-6-ylboronic acid
Inchi Code 1S/C7H7BN2O2/c11-8(12)6-1-2-7-9-3-4-10(7)5-6/h1-5,11-12H
Inchi Key IPJIKGJKMCILGV-UHFFFAOYSA-N

Description

Synonym 6-Boronoimidazo[1,2-a]pyridine
Purity 98%
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 115-117
Product Link https://store.apolloscientific.co.uk/product/imidazo12-apyridine-6-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/imidazo12-apyridine-6-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart