APO455830315 Apollo Scientific Ltd

4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid

  • MDL number MFCD08436062
  • Cas Number : 913835-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-190
Synonym 2-(4-Boronophenyl)-5-methyl-1,3,4-oxadiazole
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32889

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32889
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart