APO455830318 Apollo Scientific Ltd

2-Chloro-5-nitrobenzeneboronic acid

  • MDL number MFCD03095323
  • Cas Number : 867333-29-7
  • Molecular Weight : 201.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-238
Synonym 3-Borono-4-chloronitrobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-nitrobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-nitrobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart