APO455830319 Apollo Scientific Ltd

3-[4-(Methylsulphonyl)phenyl]propanoic acid

  • MDL number MFCD01319236
  • Cas Number : 387350-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[4-(Methylsulphonyl)phenyl]propionic acid, 4-(3-Hydroxy-3-oxoprop-1-yl)phenyl methyl sulphone
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15346

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=15346
Availability details for
Quantity
  • Availability
  • Add to Cart