APO455830324 Apollo Scientific Ltd

4-Acetoxy-2-bromo-5-methoxybenzaldehyde

  • MDL number MFCD00452230
  • Cas Number : 52783-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-113
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32909

Safety Information

signalword Warning
pcodes P280;P302+P352;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32909
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart