APO455830329 Apollo Scientific Ltd

2-Chloro-6-nitrobenzoic acid

  • MDL number MFCD00100425
  • Cas Number : 5344-49-0
  • Molecular Weight : 201.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 160-166
Iupac Name 2-chloro-6-nitrobenzoic acid
Inchi Code 1S/C7H4ClNO4/c8-4-2-1-3-5(9(12)13)6(4)7(10)11/h1-3H,(H,10,11)
Inchi Key JYHOMEFOTKWQPN-UHFFFAOYSA-N

Description

Synonym 2-Carboxy-3-chloronitrobenzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 160-166
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart