APO455830332 Apollo Scientific Ltd

6-Bromonicotinamide

  • MDL number MFCD00266840
  • Cas Number : 889676-37-3
  • Molecular Weight : 201.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-240
Synonym 6-Bromopyridine-3-carboxamide, 2-Bromo-5-carbamoylpyridine
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromonicotinamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromonicotinamide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart