APO455830336 Apollo Scientific Ltd

Ethyl imidazo[1,2-a]pyridine-7-carboxylate

  • MDL number MFCD08436046
  • Cas Number : 372147-49-4
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 87-88
Iupac Name ethyl imidazo[1,2-a]pyridine-7-carboxylate
Inchi Code 1S/C10H10N2O2/c1-2-14-10(13)8-3-5-12-6-4-11-9(12)7-8/h3-7H,2H2,1H3
Inchi Key UWYWNAGXCZHLLM-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 87-88
Product Link https://store.apolloscientific.co.uk/product/ethyl-imidazo12-apyridine-7-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-imidazo12-apyridine-7-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart