APO455830338 Apollo Scientific Ltd

4-(Methylsulphonyl)-2-nitrobenzoic acid

  • MDL number MFCD01941299
  • Cas Number : 110964-79-9
  • Molecular Weight : 245.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 211-212
Iupac Name 4-(methylsulfonyl)-2-nitrobenzoic acid
Inchi Code 1S/C8H7NO6S/c1-16(14,15)5-2-3-6(8(10)11)7(4-5)9(12)13/h2-4H,1H3,(H,10,11)
Inchi Key QNOUABMNRMROSL-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-3-nitrophenyl methyl sulphone, 2-Carboxy-5-(methylsulphonyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 211-212
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart