APO455830341 Apollo Scientific Ltd

2-(Piperazin-1-yl)-N-(pyridin-3-yl)acetamide trihydrochloride

  • MDL number MFCD03844727
  • Cas Number : 808764-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-{[(Piperazin-1-yl)acetyl]amino}pyridine trihydrochloride, 1-{2-Oxo-2-[(pyridin-3-yl)amino]ethyl}piperazine trihydrochloride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15628

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15628
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart