APO455830346 Apollo Scientific Ltd

Methyl 2-bromo-3-methoxypropanoate

  • MDL number MFCD07784248
  • Cas Number : 27704-96-7
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 87
Iupac Name methyl 2-bromo-3-methoxypropanoate
Inchi Code 1S/C5H9BrO3/c1-8-3-4(6)5(7)9-2/h4H,3H2,1-2H3
Inchi Key NVBLRKGCQVZDMQ-UHFFFAOYSA-N

Description

Melting Point 87
Synonym Methyl 2-bromo-3-methoxypropionate
Purity 95%
Flash Point 69/11mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-3-methoxypropanoate

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-3-methoxypropanoate
pictograms GHS02;GHS07
hstatements H226;H315;H319;EUH018
Availability details for
Quantity
  • Availability
  • Add to Cart