APO455830352 Apollo Scientific Ltd

3-Chloropyrazine-2-carbonitrile

  • MDL number MFCD00219653
  • Cas Number : 55557-52-3
  • Molecular Weight : 139.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 38-40
Iupac Name 3-chloro-2-pyrazinecarbonitrile
Inchi Code 1S/C5H2ClN3/c6-5-4(3-7)8-1-2-9-5/h1-2H
Inchi Key SDLFAEGTVBPHBK-UHFFFAOYSA-N

Description

Synonym 2-Chloro-3-cyanopyrazine, 2-Chloro-3-cyano-1,4-diazine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 38-40
Product Link https://store.apolloscientific.co.uk/product/3-chloropyrazine-2-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloropyrazine-2-carbonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart