APO455830361 Apollo Scientific Ltd

2-Chloro-4-(methoxycarbonyl)benzeneboronic acid

  • MDL number MFCD08689530
  • Cas Number : 603122-80-1
  • Molecular Weight : 214.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 140-142
Iupac Name 2-chloro-4-(methoxycarbonyl)phenylboronic acid
Inchi Code 1S/C8H8BClO4/c1-14-8(11)5-2-3-6(9(12)13)7(10)4-5/h2-4,12-13H,1H3
Inchi Key ITEQHBSWRMYSRP-UHFFFAOYSA-N

Description

Synonym Methyl 4-borono-3-chlorobenzoate
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 140-142
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-methoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-methoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart