APO455830366 Apollo Scientific Ltd

4-Chloro-2-formylbenzeneboronic acid

  • MDL number MFCD08689492
  • Cas Number : 913835-76-4
  • Molecular Weight : 184.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 132-134
Iupac Name 4-chloro-2-formylphenylboronic acid
Inchi Code 1S/C7H6BClO3/c9-6-1-2-7(8(11)12)5(3-6)4-10/h1-4,11-12H
Inchi Key BTQFQGHQBOWNAJ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 132-134
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-formylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-formylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart