APO455830368 Apollo Scientific Ltd

4-[(Benzylcarbamoyl)amino]benzeneboronic acid, pinacol ester

  • MDL number MFCD08689513
  • Cas Number : 874290-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-207
Synonym 4-(3-Benzylureido)benzeneboronic acid, pinacol ester
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32956

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32956
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart