APO455830372 Apollo Scientific Ltd

4-[(Cyclopropylcarbamoyl)amino]benzeneboronic acid, pinacol ester

  • MDL number MFCD08689493
  • Cas Number : 874297-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-229
Synonym 4-(3-Cyclopropylureido)benzeneboronic acid, pinacol ester
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33482

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33482
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart