APO455830384 Apollo Scientific Ltd

3-Bromo-5-chlorotoluene

  • MDL number MFCD08437596
  • Cas Number : 329944-72-1
  • Molecular Weight : 205.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-3-chloro-5-methylbenzene
Inchi Code 1S/C7H6BrCl/c1-5-2-6(8)4-7(9)3-5/h2-4H,1H3
Inchi Key YRIKDGJWRMHTJP-UHFFFAOYSA-N

Description

Synonym 1-Bromo-3-chloro-5-methylbenzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-chlorotoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-chlorotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart