APO455830385 Apollo Scientific Ltd

6-Chloro-5-nitronicotinic acid

  • MDL number MFCD09027297
  • Cas Number : 7477-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-102
Synonym 6-Chloro-5-nitropyridine-3-carboxylic acid, 5-Carboxy-2-chloro-3-nitropyridine
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36390

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36390
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart