APO455830386 Apollo Scientific Ltd

4-[(4-Chlorophenyl)carbamoyl]benzeneboronic acid

  • MDL number MFCD09027203
  • Cas Number : 874288-02-5
  • Molecular Weight : 275.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 248-250
Iupac Name 4-[(4-chloroanilino)carbonyl]phenylboronic acid
Inchi Code 1S/C13H11BClNO3/c15-11-5-7-12(8-6-11)16-13(17)9-1-3-10(4-2-9)14(18)19/h1-8,18-19H,(H,16,17)
Inchi Key KQGSKPWDDNXYNQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 248-250
Product Link https://store.apolloscientific.co.uk/product/4-4-chlorophenylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-4-chlorophenylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart