APO455830389 Apollo Scientific Ltd

4-[2-(Piperidin-1-yl)ethoxy]benzeneboronic acid, pinacol ester

  • MDL number MFCD09258652
  • Cas Number : 934586-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-{2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]ethyl}piperidine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-2-piperidin-1-ylethoxybenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-2-piperidin-1-ylethoxybenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H302;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart