APO455830406 Apollo Scientific Ltd

Methyl 5-methyl-1,3-oxazole-4-carboxylate

  • MDL number MFCD08689502
  • Cas Number : 41172-57-0
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 45-48
Iupac Name methyl 5-methyl-1,3-oxazole-4-carboxylate
Inchi Code 1S/C6H7NO3/c1-4-5(6(8)9-2)7-3-10-4/h3H,1-2H3
Inchi Key UMYOVEXLUCPUAU-UHFFFAOYSA-N

Description

Synonym 4-(Methoxycarbonyl)-5-methyl-1,3-oxazole
Purity 97%
Flash Point 51-57/0.1mm
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 45-48
Product Link https://store.apolloscientific.co.uk/product/methyl-5-methyl-13-oxazole-4-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-methyl-13-oxazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart