APO455830407 Apollo Scientific Ltd

N,N-Dimethylindoline-5-sulphonamide

  • MDL number MFCD00214476
  • Cas Number : 99169-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-111
Synonym 5-(N,N-Dimethylsulphamoyl)indoline, 2,3-Dihydro-N,N-dimethyl-1H-indole-5-sulphonamide, 2,3-Dihydro-5-(N,N-dimethylsulphamoyl)-1H-indole
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14307

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14307
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart