APO455830412 Apollo Scientific Ltd

2-Methoxy-5-sulphamoylbenzoic acid

  • MDL number MFCD00129997
  • Cas Number : 22117-85-7
  • Molecular Weight : 231.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(aminosulfonyl)-2-methoxybenzoic acid
Inchi Code 1S/C8H9NO5S/c1-14-7-3-2-5(15(9,12)13)4-6(7)8(10)11/h2-4H,1H3,(H,10,11)(H2,9,12,13)
Inchi Key SQAILWDRVDGLGY-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-4-methoxybenzenesulphonamide, 5-(Aminosulphonyl)-o-anisic acid
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-5-sulphamoylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-5-sulphamoylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart