APO455830416 Apollo Scientific Ltd

Methyl 5-(ethylsulphonyl)-2-methoxybenzoate

  • MDL number MFCD01317547
  • Cas Number : 62140-67-4
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 126
Iupac Name methyl 5-(ethylsulfonyl)-2-methoxybenzoate
Inchi Code 1S/C11H14O5S/c1-4-17(13,14)8-5-6-10(15-2)9(7-8)11(12)16-3/h5-7H,4H2,1-3H3
Inchi Key UUARTHQJSZTOEM-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 126
Product Link https://store.apolloscientific.co.uk/product/methyl-5-ethylsulphonyl-2-methoxybenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-ethylsulphonyl-2-methoxybenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart