APO455830417 Apollo Scientific Ltd

Methyl 4-amino-5-(ethylsulphonyl)-2-methoxybenzoate

  • MDL number MFCD07787259
  • Cas Number : 80036-89-1
  • Molecular Weight : 273.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-amino-5-(ethylsulfonyl)-2-methoxybenzoate
Inchi Code 1S/C11H15NO5S/c1-4-18(14,15)10-5-7(11(13)17-3)9(16-2)6-8(10)12/h5-6H,4,12H2,1-3H3
Inchi Key BBWJVKZAKHIKRQ-UHFFFAOYSA-N

Description

Synonym 2-(Ethylsulphonyl)-5-methoxy-4-(methoxycarbonyl)aniline, 5-Amino-4-(ethylsulphonyl)-2-(methoxycarbonyl)anisole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-amino-5-ethylsulphonyl-2-methoxybenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-amino-5-ethylsulphonyl-2-methoxybenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart