APO455830419 Apollo Scientific Ltd

2-Bromo-5-methoxypyridine

  • MDL number MFCD07781221
  • Cas Number : 105170-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-methoxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-methoxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart