APO455830421 Apollo Scientific Ltd

6-Chloro-2-methyl-3-nitropyridine

  • MDL number MFCD00234160
  • Cas Number : 22280-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-58
Synonym 6-Chloro-3-nitro-2-picoline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-chloro-2-methyl-3-nitropyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-2-methyl-3-nitropyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart