APO455830425 Apollo Scientific Ltd

3-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid

  • MDL number MFCD08689540
  • Cas Number : 913836-04-1
  • Molecular Weight : 203.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-162
Synonym 2-(3-Boronophenyl)-5-methyl-1,3,4-oxadiazole
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-5-methyl-134-oxadiazol-2-ylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-5-methyl-134-oxadiazol-2-ylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart