APO455830430 Apollo Scientific Ltd

5-(Methylsulphonyl)pyridine-3-boronic acid

  • MDL number MFCD08689536
  • Cas Number : 913836-01-8
  • Molecular Weight : 201.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 123-125
Iupac Name 5-(methylsulfonyl)-3-pyridinylboronic acid
Inchi Code 1S/C6H8BNO4S/c1-13(11,12)6-2-5(7(9)10)3-8-4-6/h2-4,9-10H,1H3
Inchi Key YGXNPQDXWDCWET-UHFFFAOYSA-N

Description

Synonym 3-Borono-5-(methylsulphonyl)pyridine, 5-Boronopyridin-3-yl methyl sulphone
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 123-125
Product Link https://store.apolloscientific.co.uk/product/5-methylsulphonylpyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methylsulphonylpyridine-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart