APO455830432 Apollo Scientific Ltd

3-Bromo-5-(methylthio)pyridine

  • MDL number MFCD08689537
  • Cas Number : 142137-18-6
  • Molecular Weight : 204.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-(methylsulfanyl)pyridine
Inchi Code 1S/C6H6BrNS/c1-9-6-2-5(7)3-8-4-6/h2-4H,1H3
Inchi Key AKFNVEXSYXJWQX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-methylthiopyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-methylthiopyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart