APO455830435 Apollo Scientific Ltd

6-Bromoisatin

  • MDL number MFCD01631138
  • Cas Number : 6326-79-0
  • Molecular Weight : 226.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 274
Iupac Name 6-bromo-1H-indole-2,3-dione
Inchi Code 1S/C8H4BrNO2/c9-4-1-2-5-6(3-4)10-8(12)7(5)11/h1-3H,(H,10,11,12)
Inchi Key HVPQMLZLINVIHW-UHFFFAOYSA-N

Description

Synonym 6-Bromo-1H-indole-2,3-dione, 6-Bromoindolin-2,3-dione
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 274
Product Link https://store.apolloscientific.co.uk/product/6-bromoisatin

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromoisatin
pictograms GHS07
hstatements H302+H312+H332;H317;H319
Availability details for
Quantity
  • Availability
  • Add to Cart