APO455830438 Apollo Scientific Ltd

2,1,3-Benzoxadiazole-5-carbonyl chloride

  • MDL number MFCD00276978
  • Cas Number : 126147-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-58
Synonym Benzofurazan-5-carbonyl chloride, 5-(Chlorocarbonyl)-2,1,3-benzoxadiazole, 5-(Chloroformyl)-2,1,3-benzoxadiazole
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12929

Safety Information

signalword Danger
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12929
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart