APO455830439 Apollo Scientific Ltd

1-Aminoindane hydrochloride

  • MDL number MFCD00054294
  • Cas Number : 70146-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-210
Synonym 2,3-Dihydro-1H-inden-1-amine hydrochloride, Indan-1-amine hydrochloride
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-aminoindane-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-aminoindane-hydrochloride
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart