APO455830444 Apollo Scientific Ltd

3-Chloro-5-methylbenzeneboronic acid

  • MDL number MFCD09027241
  • Cas Number : 913836-14-3
  • Molecular Weight : 170.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 235-238
Iupac Name 3-chloro-5-methylphenylboronic acid
Inchi Code 1S/C7H8BClO2/c1-5-2-6(8(10)11)4-7(9)3-5/h2-4,10-11H,1H3
Inchi Key QXHLBINWDJITMJ-UHFFFAOYSA-N

Description

Synonym 3-Borono-5-chlorotoluene
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 235-238
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-methylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-methylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart