APO455830445 Apollo Scientific Ltd

6-Bromo-2-iminocoumarin-3-carbonitrile

  • MDL number MFCD00115365
  • Cas Number : 860789-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-3-cyano-2-iminocoumarin, 6-Bromo-2-imino-2H-chromene-3-carbonitrile
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16032

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16032
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart