APO455830447 Apollo Scientific Ltd

3-Carboxy-4-methoxybenzeneboronic acid

  • MDL number MFCD06203322
  • Cas Number : 913836-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-177
Synonym 5-Borono-2-methoxybenzoic acid, 4-Borono-2-carboxyanisole
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=35781

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=35781
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart