APO455830468 Apollo Scientific Ltd

1-Acetyl-2-methylindoline-5-sulphonyl chloride

  • MDL number MFCD07841689
  • Cas Number : 841275-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Synonym 1-Acetyl-5-(chlorosulphonyl)-2-methylindoline, 1-Acetyl-2,3-dihydro-2-methyl-1H-indole-5-sulphonyl chloride
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=35941

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=35941
pictograms GHS05
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart